siRNA Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291765.1 | NP_001278694.1 | 9 | 885 | ||
| NM_001291766.1 | NP_001278695.1 | 8 | 879 | |||
| NM_001291768.1 | NP_001278697.1 | 8 | 818 | |||
| NM_009361.3 | NP_033387.1 | 9 | 946 | |||
| GenBank mRNA | AK013180.1 | - | 8 | 848 | ||
| AK019997.1 | - | 9 | 886 | |||
| AK030820.1 | - | 9 | 947 | |||
| AK154037.1 | - | 8 | 786 | |||
| AK198738.1 | - | 3 | 271 | |||
| BC058633.1 | - | 8 | 1161 | |||
| BC132567.1 | - | 8 | 791 | 40130844 | ||
| BC132569.1 | - | 8 | 791 | 40130846 | ||
| BC145344.1 | - | 8 | 791 | 9053900 | ||
| X72310.1 | - | 8 | 820 | |||

Tfdp1
Genomic Map