siRNA Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252443.1 | NP_001239372.1 | 3 | 549 | ||
| NM_019678.3 | NP_062652.1 | 3 | 549 | |||
| GenBank mRNA | AK008549.1 | - | 3 | 325 | ||
| AK088864.1 | - | 3 | 367 | |||
| AK158539.1 | - | 3 | 415 | |||
| AK167475.1 | - | 3 | 413 | |||
| AK186338.1 | - | 2 | 287 | |||
| AK186588.1 | - | 2 | 199 | |||
| AK210251.1 | - | 2 | 198 | |||
| BC024638.1 | - | 3 | 327 | 4015300 | ||
| U94662.1 | - | 3 | 451 | |||

Tfg
Genomic Map