siRNA Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019678.3 | NP_062652.1 | 4 | 647 | ||
| GenBank mRNA | AK008549.1 | - | 4 | 423 | ||
| AK158539.1 | - | 4 | 513 | |||
| AK167475.1 | - | 4 | 511 | |||
| BC024638.1 | - | 4 | 425 | 4015300 | ||
| U94662.1 | - | 4 | 549 | |||

Tfg
Genomic Map