siRNA Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289550.1 | NP_001276479.1 | 7 | 879 | ||
| NM_001289551.1 | NP_001276480.1 | 7 | 879 | |||
| NM_001289552.1 | NP_001276481.1 | 6 | 736 | |||
| NM_001289553.1 | NP_001276482.1 | 6 | 763 | |||
| GenBank mRNA | AK003170.1 | - | 7 | 713 | ||
| AK041233.1 | - | 7 | 708 | |||
| AK085042.1 | - | 6 | 605 | |||
| AK087114.1 | - | 7 | 714 | |||
| AK156423.1 | - | 7 | 714 | |||
| AK158409.1 | - | 6 | 588 | |||
| BC002049.1 | - | 7 | 713 | 3489459 | ||
| BC056362.1 | - | 6 | 1988 | 6816930 | ||
| DQ143891.1 | - | 6 | 764 | |||
| DQ143892.1 | - | 5 | 550 | |||
| DQ143893.1 | - | 5 | 557 | |||
| DQ143894.1 | - | 5 | 552 | |||
| DQ143895.1 | - | 5 | 968 | |||
| DQ143896.1 | - | 5 | 929 | |||
| DQ143897.1 | - | 5 | 1007 | |||
| DQ143898.1 | - | 5 | 956 | |||
| DQ143899.1 | - | 4 | 1083 | |||
| DQ143900.1 | - | 6 | 776 | |||
| L22482.1 | - | 6 | 840 | |||

Tgfb1i1
Genomic Map