siRNA Details
| Gene Symbol: Tgfbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009370.2 | NP_033396.1 | 7 | 1203 | ||
| GenBank mRNA | AK007125.1 | - | 3 | 334 | ||
| AK015337.1 | - | 6 | 1139 | |||
| AK154381.1 | - | 6 | 966 | |||
| BC063260.1 | - | 7 | 1203 | 6409765 | ||
| CT010393.1 | - | 7 | 1196 | |||
| D25540.1 | - | 7 | 1189 | |||
| D28526.1 | - | 7 | 1196 | |||

Tgfbr1
Genomic Map