siRNA Details
| Gene Symbol: Tg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009375.2 | NP_033401.2 | 7 | 866 | ||
| GenBank mRNA | AF076186.1 | - | 7 | 844 | ||
| AF076187.1 | - | 7 | 844 | |||
| BC111467.1 | - | 7 | 859 | 30841130 | ||
| U76389.1 | - | 7 | 883 | |||

Tg
Genomic Map