siRNA Details
| Gene Symbol: Ift88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009376.2 | NP_033402.2 | 26 | 2551 | ||
| GenBank mRNA | AK135110.1 | - | 26 | 2546 | ||
| AK168395.1 | - | 23 | 3699 | |||
| BC012250.1 | - | 26 | 2531 | 4019334 | ||
| L31959.1 | - | 26 | 2496 | |||

Ift88
Genomic Map