siRNA Details
| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009377.1 | NP_033403.1 | 13 | 1383 | ||
| GenBank mRNA | AK139568.1 | - | 1600 | |||
| M69200.1 | - | 13 | 1383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Th| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009377.1 | NP_033403.1 | 13 | 1383 | ||
| GenBank mRNA | AK139568.1 | - | 1600 | |||
| M69200.1 | - | 13 | 1383 | |||