siRNA Details
| Gene Symbol: Thbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011581.3 | NP_035711.2 | 16 | 2831 | ||
| GenBank mRNA | AK132508.1 | - | 16 | 2662 | ||
| AK142552.1 | - | 16 | 2803 | |||
| BC031843.1 | - | 5 | 616 | |||
| BC053702.1 | - | 16 | 2633 | 30062369 | ||
| L07803.1 | - | 16 | 2659 | |||
| M64866.1 | - | 16 | 2659 | |||

Thbs2
Genomic Map