siRNA Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013691.2 | NP_038719.2 | 17 | 2106 | ||
| GenBank mRNA | BC053023.1 | - | 17 | 2060 | 6401682 | |
| BC060104.1 | - | 15 | 3098 | |||
| L04302.1 | - | 17 | 2086 | |||
| L24434.1 | - | 17 | 2083 | |||

Thbs3
Genomic Map