siRNA Details
| Gene Symbol: Thbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011582.3 | NP_035712.1 | 14 | 1932 | ||
| GenBank mRNA | AF102887.1 | - | 14 | 1872 | ||
| BC139414.1 | - | 14 | 1812 | 8862436 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thbs4| Gene Symbol: Thbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011582.3 | NP_035712.1 | 14 | 1932 | ||
| GenBank mRNA | AF102887.1 | - | 14 | 1872 | ||
| BC139414.1 | - | 14 | 1812 | 8862436 | ||