siRNA Details
| Gene Symbol: Theg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011583.3 | NP_035713.1 | 8 | 1149 | ||
| NM_199226.1 | NP_954696.1 | 7 | 1077 | |||
| GenBank mRNA | AB033128.1 | - | 8 | 1131 | ||
| AJ011834.1 | - | 8 | 1337 | |||
| AK017058.1 | - | 7 | 1067 | |||
| BC050970.1 | - | 7 | 1078 | |||
| BC061243.1 | - | 7 | 1023 | 6775002 | ||

Theg
Genomic Map