siRNA Details
| Gene Symbol: Thpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173505.1 | NP_001166976.1 | 6 | 816 | ||
| NM_001289894.1 | NP_001276823.1 | 6 | 804 | |||
| NM_009379.3 | NP_033405.1 | 6 | 970 | |||
| GenBank mRNA | AK047512.1 | - | 6 | 971 | ||
| BC003803.1 | - | 6 | 1082 | 3593885 | ||
| L34169.1 | - | 6 | 752 | |||

Thpo
Genomic Map