siRNA Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113417.1 | NP_001106888.1 | 10 | 1410 | ||
| NM_009380.3 | NP_033406.1 | 7 | 1496 | |||
| GenBank mRNA | AB221613.1 | - | 7 | 1263 | ||
| AK133662.1 | - | 7 | 993 | |||
| AK158826.1 | - | 10 | 1411 | |||
| BC089035.1 | - | 7 | 1496 | 30695885 | ||
| BC119552.1 | - | 7 | 1433 | 40057420 | ||
| BC119553.1 | - | 7 | 1437 | 40057421 | ||
| S62756.1 | - | 9 | 1280 | |||

Thrb
Genomic Map