siRNA Details
| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009381.2 | NP_033407.1 | 2 | 819 | ||
| GenBank mRNA | AK077300.1 | - | 2 | 612 | ||
| BC009165.1 | - | 2 | 819 | 2811774 | ||
| BC053378.1 | - | 1 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thrsp| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009381.2 | NP_033407.1 | 2 | 819 | ||
| GenBank mRNA | AK077300.1 | - | 2 | 612 | ||
| BC009165.1 | - | 2 | 819 | 2811774 | ||
| BC053378.1 | - | 1 | 270 | |||