siRNA Details
| Gene Symbol: Tia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164078.1 | NP_001157550.1 | 10 | 1116 | ||
| NM_011585.4 | NP_035715.1 | 11 | 1143 | |||
| GenBank mRNA | AK028340.1 | - | 9 | 783 | ||
| AK088486.1 | - | 11 | 1079 | |||
| AK144222.1 | - | 11 | 1096 | |||
| AK190403.1 | - | 2 | 201 | |||
| BC027826.1 | - | 9 | 3080 | |||
| BC046812.1 | - | 10 | 999 | 5702183 | ||
| U00689.1 | - | 11 | 1084 | |||
| U55862.1 | - | 11 | 951 | |||

Tia1
Genomic Map