siRNA Details
| Gene Symbol: Timm17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011591.4 | NP_035721.1 | 3, 4 | 241 | ||
| GenBank mRNA | AF106621.1 | - | 3, 4 | 195 | ||
| AK003204.1 | - | 3, 4 | 214 | |||
| AK009164.1 | - | 3, 4 | 243 | |||
| AK011830.1 | - | 3, 4 | 242 | |||
| AK049584.1 | - | 3, 4 | 242 | |||
| AK150836.1 | - | 3, 4 | 244 | |||
| AK153097.1 | - | 3, 4 | 242 | |||
| BC008275.1 | - | 3, 4 | 223 | 2811777 | ||

Timm17b
Genomic Map