siRNA Details
| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009388.6 | NP_033414.1 | 2 | 346 | ||
| GenBank mRNA | AF195533.1 | - | 1 | 12 | ||
| AK002627.1 | - | 2 | 299 | |||
| AK012794.1 | - | 2 | 301 | |||
| AK030446.1 | - | 2 | 329 | |||
| AK050793.1 | - | 2 | 297 | |||
| AK140965.1 | - | 2 | 330 | |||
| AK144146.1 | - | 2 | 301 | |||
| AK145677.1 | - | 2 | 299 | |||
| AK146157.1 | - | 2 | 299 | |||
| AK150139.1 | - | 2 | 299 | |||
| AK150769.1 | - | 2 | 295 | |||
| AK150844.1 | - | 2 | 295 | |||
| AK150856.1 | - | 2 | 295 | |||
| AK150901.1 | - | 2 | 299 | |||
| AK151916.1 | - | 2 | 295 | |||
| AK151969.1 | - | 2 | 295 | |||
| AK152245.1 | - | 2 | 295 | |||
| AK152460.1 | - | 2 | 315 | |||
| AK152886.1 | - | 2 | 295 | |||
| AK159922.1 | - | 2 | 299 | |||
| AK166763.1 | - | 2 | 333 | |||
| AK167084.1 | - | 2 | 333 | |||
| AK181952.1 | - | 1 | 111 | |||
| BC055336.1 | - | 2 | 273 | 6413263 | ||
| BY276711.1 | - | 2 | 347 | |||

Tkt
Genomic Map