siRNA Details
| Gene Symbol: Tln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011602.5 | NP_035732.2 | 11 | 1621 | ||
| GenBank mRNA | AK040128.1 | - | 8 | 837 | ||
| AK122420.2 | - | 11 | 1247 | |||
| AK147228.1 | - | 11 | 1192 | |||
| BC150810.1 | - | 11 | 1292 | 9087721 | ||
| X56123.1 | - | 11 | 1293 | |||

Tln1
Genomic Map