siRNA Details
| Gene Symbol: Otop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172709.3 | NP_766297.2 | 6 | 1741 | ||
| GenBank mRNA | AF548337.1 | - | 6 | 1741 | ||
| AK040792.1 | - | 6 | 1728 | |||
| AK041023.1 | - | 6 | 1515 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_181433.

Otop1
Genomic Map