siRNA Details
Gene Symbol: Tlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009392.2 | NP_033418.1 | 3 | 1217 | ||
GenBank mRNA | AK013072.1 | - | 2 | 1293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009392.2 | NP_033418.1 | 3 | 1217 | ||
GenBank mRNA | AK013072.1 | - | 2 | 1293 |