siRNA Details
| Gene Symbol: Tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020275.4 | NP_064671.2 | 3 | 459 | ||
| GenBank mRNA | AB031081.1 | - | 3 | 399 | ||
| AF176833.1 | - | 3 | 271 | |||
| AK050753.1 | - | 3 | 460 | |||
| AK080069.1 | - | 3 | 450 | |||
| AK145730.1 | - | 3 | 447 | |||
| AK154993.1 | - | 3 | 439 | |||
| BC065141.1 | - | 3 | 402 | 30608306 | ||

Tnfrsf10b
Genomic Map