siRNA Details
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033126.2 | NP_001028298.1 | 6 | 841 | ||
| NM_001042564.1 | NP_001036029.1 | 4 | 529 | |||
| NM_001286753.1 | NP_001273682.1 | 5 | 661 | |||
| GenBank mRNA | AK153895.1 | - | 6 | 780 | ||
| AK154005.1 | - | 6 | 825 | |||
| BC139125.1 | - | 6 | 806 | 8862147 | ||
| BC139126.1 | - | 6 | 806 | 8862148 | ||
| BC171931.1 | - | 4 | 494 | 9054107 | ||
| BE631582.1 | - | 5 | 480 | |||
| L24495.1 | - | 6 | 883 | |||

Cd27
Genomic Map