siRNA Details
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033126.2 | NP_001028298.1 | 5 | 738 | ||
| NM_001042564.1 | NP_001036029.1 | 3 | 426 | |||
| NM_001286753.1 | NP_001273682.1 | 4 | 558 | |||
| GenBank mRNA | AK153895.1 | - | 5 | 677 | ||
| AK154005.1 | - | 5 | 722 | |||
| BC139125.1 | - | 5 | 703 | 8862147 | ||
| BC139126.1 | - | 5 | 703 | 8862148 | ||
| BC171931.1 | - | 3 | 391 | 9054107 | ||
| BE631582.1 | - | 4 | 377 | |||
| DT924276.1 | - | 4 | 484 | |||
| L24495.1 | - | 5 | 780 | |||

Cd27
Genomic Map