siRNA Details
| Gene Symbol: Tnfrsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077508.1 | NP_001070976.1 | 3, 4 | 345 | ||
| NM_001077509.1 | NP_001070977.1 | 3, 4 | 326 | |||
| NM_011612.2 | NP_035742.1 | 3, 4 | 345 | |||
| GenBank mRNA | AK019885.1 | - | 3, 4 | 311 | ||
| AK154625.1 | - | 3, 4 | 329 | |||
| BC028507.1 | - | 3, 4 | 282 | 1497753 | ||
| BY206772.1 | - | 3, 4 | 345 | |||
| DQ832278.1 | - | 2, 3 | 231 | |||
| DQ832279.1 | - | 2, 3 | 231 | |||

Tnfrsf9
Genomic Map