siRNA Details
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130174.2 | NP_001123646.1 | 14, 15 | 943 | ||
NM_001130175.2 | NP_001123647.4 | 14, 15 | 939 | |||
NM_001130176.1 | NP_001123648.1 | 13, 14 | 839 | |||
NM_001130177.2 | NP_001123649.1 | 13, 14 | 931 | |||
NM_001130178.2 | NP_001123650.1 | 14, 15 | 922 | |||
NM_001130179.2 | NP_001123651.4 | 14, 15 | 918 | |||
NM_001130180.2 | NP_001123652.1 | 13, 14 | 910 | |||
NM_001130181.2 | NP_001123653.4 | 13, 14 | 906 | |||
NM_011619.3 | NP_035749.1 | 12, 13 | 901 | |||
GenBank mRNA | AK146945.1 | - | 14, 15 | 888 | ||
AK168393.1 | - | 13, 14 | 876 | |||
AK169061.1 | - | 14, 15 | 868 | |||
BC063753.1 | - | 14, 15 | 919 | 30293959 | ||
L47549.1 | - | 14, 15 | 836 | |||
L47550.1 | - | 14, 15 | 821 | |||
L47551.1 | - | 13, 14 | 822 | |||
L47552.1 | - | 13, 14 | 831 | |||
L47553.1 | - | 13, 14 | 838 | |||
L47599.1 | - | 14, 15 | 842 | |||
L47600.1 | - | 12, 13 | 830 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
