siRNA Details
| Gene Symbol: ITGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181501.1 | NP_852478.1 | 18 | 2391 | ||
| GenBank mRNA | AK300423.1 | - | 18 | 2556 | ||
| BC137121.1 | - | 18 | 2390 | 9021118 | ||
| BC137122.1 | - | 18 | 2390 | 9021119 | ||
| BX640981.1 | - | 7 | 5014 | |||
| X68742.1 | - | 17 | 2255 | |||

ITGA1
Genomic Map