siRNA Details
| Gene Symbol: Tnnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163664.1 | NP_001157136.1 | 15 | 680 | ||
| NM_001163665.1 | NP_001157137.1 | 15 | 680 | |||
| NM_001163666.1 | NP_001157138.1 | 13 | 653 | |||
| NM_001163667.1 | NP_001157139.1 | 14 | 668 | |||
| NM_001163668.1 | NP_001157140.1 | 13 | 629 | |||
| NM_001163669.1 | NP_001157141.1 | 14 | 641 | |||
| NM_001163670.1 | NP_001157142.1 | 11 | 596 | |||
| NM_011620.3 | NP_035750.2 | 12 | 608 | |||
| GenBank mRNA | AI746973.1 | - | 13 | 648 | ||
| AK003147.1 | - | 11 | 563 | |||
| BC003747.1 | - | 15 | 676 | 3594256 | ||
| CB196352.1 | - | 5 | 432 | |||
| CK791812.1 | - | 4 | 318 | |||
| DV049834.1 | - | 12 | 655 | |||
| L48988.1 | - | 9 | 499 | |||
| L48989.1 | - | 9 | 499 | |||
| L48990.1 | - | 12 | 544 | |||
| L48991.1 | - | 9 | 526 | |||
| L48992.1 | - | 11 | 532 | |||
| L48993.1 | - | 11 | 532 | |||
| L49018.1 | - | 9 | 526 | |||
| L49466.1 | - | 14 | 668 | |||
| L49467.1 | - | 15 | 680 | |||
| L49468.1 | - | 13 | 653 | |||
| L49469.1 | - | 13 | 653 | |||
| L49470.1 | - | 14 | 641 | |||
| L49471.1 | - | 13 | 650 | |||
| L49472.1 | - | 13 | 629 | |||
| U77779.1 | - | 12 | 601 | |||

Tnnt3
Genomic Map