siRNA Details
| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009409.2 | NP_033435.2 | 30 | 4407 | ||
| GenBank mRNA | BC041106.1 | - | 30 | 4427 | 5368508 | |
| BC054541.1 | - | 30 | 4407 | 6406462 | ||
| D38046.1 | - | 30 | 3980 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Top2b| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009409.2 | NP_033435.2 | 30 | 4407 | ||
| GenBank mRNA | BC041106.1 | - | 30 | 4427 | 5368508 | |
| BC054541.1 | - | 30 | 4407 | 6406462 | ||
| D38046.1 | - | 30 | 3980 | |||