siRNA Details
| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011626.2 | NP_035756.2 | 5 | 1131 | ||
| GenBank mRNA | BC037638.1 | - | 6 | 912 | ||
| BC141081.1 | - | 5 | 915 | 9055693 | ||
| M23568.1 | - | 5 | 1062 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem165| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011626.2 | NP_035756.2 | 5 | 1131 | ||
| GenBank mRNA | BC037638.1 | - | 6 | 912 | ||
| BC141081.1 | - | 5 | 915 | 9055693 | ||
| M23568.1 | - | 5 | 1062 | |||