siRNA Details
| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009413.1 | NP_033439.1 | 3 | 351 | ||
| GenBank mRNA | AF004428.1 | - | 3 | 351 | ||
| AK131855.1 | - | 3 | 248 | |||
| BC141068.1 | - | 3 | 238 | 9055680 | ||
| BC145321.1 | - | 3 | 238 | 9053876 | ||
| BC145322.1 | - | 3 | 238 | |||
| BC171924.1 | - | 3 | 238 | |||

Tpd52l1
Genomic Map