siRNA Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016785.2 | NP_058065.2 | 9 | 805 | ||
| GenBank mRNA | AF037043.1 | - | 9 | 714 | ||
| AF037044.1 | - | 8 | 665 | |||
| AF037045.1 | - | 8 | 665 | |||
| AF046887.1 | - | 9 | 791 | |||
| AK079074.1 | - | 2 | 60 | |||
| BC021598.1 | - | 8 | 692 | 5098857 | ||

Tpmt
Genomic Map