siRNA Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009417.2 | NP_033443.1 | 13 | 2286 | ||
| GenBank mRNA | AK047843.1 | - | 13 | 2265 | ||
| X60703.1 | - | 13 | 2286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpo| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009417.2 | NP_033443.1 | 13 | 2286 | ||
| GenBank mRNA | AK047843.1 | - | 13 | 2265 | ||
| X60703.1 | - | 13 | 2286 | |||