siRNA Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204071.1 | NP_001191000.1 | 9, 10 | 854 | ||
| NM_009420.2 | NP_033446.1 | 9, 10 | 872 | |||
| GenBank mRNA | BC049615.1 | - | 9, 10 | 824 | 6742033 | |
| M25533.1 | - | 9, 10 | 824 | |||

Crisp2
Genomic Map