siRNA Details
| Gene Symbol: Traf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290413.1 | NP_001277342.1 | 7, 8 | 810 | ||
| NM_009422.3 | NP_033448.2 | 7, 8 | 789 | |||
| GenBank mRNA | AF027570.1 | - | 6, 7 | 701 | ||
| AK038136.1 | - | 7, 8 | 738 | |||
| AK052934.1 | - | 7, 8 | 738 | |||
| AK151653.1 | - | 7, 8 | 727 | |||
| AK152174.1 | - | 7, 8 | 757 | |||
| AK159581.1 | - | 7, 8 | 759 | |||
| AK159585.1 | - | 736 | ||||
| BC003801.1 | - | 6, 7 | 715 | 3492839 | ||
| BC060625.1 | - | 5, 6 | 2746 | |||
| L35303.1 | - | 7, 8 | 726 | |||

Traf2
Genomic Map