siRNA Details
| Gene Symbol: Traf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011633.2 | NP_035763.2 | 964 | |||
| GenBank mRNA | AK141097.1 | - | 6, 7 | 925 | ||
| AK144694.1 | - | 915 | ||||
| BC012702.1 | - | 918 | 4218625 | |||
| D78141.1 | - | 941 | ||||
| D83528.1 | - | 806 | ||||

Traf5
Genomic Map