siRNA Details
| Gene Symbol: Trex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012236.1 | NP_001012236.1 | 2 | 482 | ||
| NM_011637.6 | NP_035767.4 | 2 | 496 | |||
| GenBank mRNA | AF151106.3 | - | 2 | 482 | ||
| AF319574.1 | - | 1 | 1196 | |||
| AK009899.1 | - | 2 | 483 | |||
| AK087832.1 | - | 2 | 483 | |||
| AK171916.1 | - | 2 | 471 | |||
| BC011133.1 | - | 2 | 503 | 4206812 | ||
| CA323364.1 | - | 2 | 636 | |||

Trex1
Genomic Map