siRNA Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133977.2 | NP_598738.1 | 16 | 2037 | ||
| GenBank mRNA | AF440692.1 | - | 15 | 1967 | ||
| AK085754.1 | - | 16 | 2022 | |||
| AK142599.1 | - | 2027 | ||||
| AK146549.1 | - | 16 | 2022 | |||
| AK149559.1 | - | 16 | 2022 | |||
| AK149595.1 | - | 16 | 2022 | |||
| AK150782.1 | - | 16 | 2070 | |||
| AK168368.1 | - | 16 | 2024 | |||
| AK168405.1 | - | 16 | 2024 | |||
| AK168419.1 | - | 16 | 2022 | |||
| AK168938.1 | - | 16 | 2022 | |||
| AK169187.1 | - | 16 | 2021 | |||
| BC008559.1 | - | 14 | 1759 | |||
| BC012313.1 | - | 16 | 2037 | 3981648 | ||
| BC018573.1 | - | 11 | 1307 | |||
| BC020295.1 | - | 16 | 2006 | |||
| BC022986.1 | - | 16 | 2015 | 5123741 | ||
| BC058216.1 | - | 16 | 2003 | |||
| BC058218.1 | - | 16 | 2016 | |||
| BC092046.1 | - | 16 | 1998 | 3594705 | ||

Trf
Genomic Map