siRNA Details
| Gene Symbol: Tob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009427.2 | NP_033453.2 | 2 | 802 | ||
| GenBank mRNA | AK142667.1 | - | 2 | 818 | ||
| BC082588.1 | - | 2 | 802 | 30629144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tob1| Gene Symbol: Tob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009427.2 | NP_033453.2 | 2 | 802 | ||
| GenBank mRNA | AK142667.1 | - | 2 | 818 | ||
| BC082588.1 | - | 2 | 802 | 30629144 | ||