siRNA Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011643.2 | NP_035773.1 | 4 | 981 | ||
| GenBank mRNA | AF191551.1 | - | 3 | 427 | ||
| BC137581.1 | - | 4 | 967 | 8860600 | ||
| BC144726.1 | - | 3 | 865 | 9053274 | ||
| U73625.1 | - | 4 | 981 | |||
| U95167.1 | - | 3 | 879 | |||
| U95168.1 | - | 3 | 826 | |||
| U95169.1 | - | 2 | 724 | |||

Trpc1
Genomic Map