siRNA Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019510.2 | NP_062383.2 | 10, 11 | 2702 | ||
| GenBank mRNA | AF190645.1 | - | 9, 10 | 2292 | ||
| AK032317.1 | - | 10, 11 | 2703 | |||
| AK080619.1 | - | 10, 11 | 2690 | |||
| BC108988.2 | - | 9, 10 | 2291 | |||
| BC108989.2 | - | 9, 10 | 2292 | 40047766 | ||
| FJ207475.1 | - | 9, 10 | 2292 | |||

Trpc3
Genomic Map