siRNA Details
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113404.1 | NP_001106875.1 | 7 | 680 | ||
NM_011648.5 | NP_035778.3 | 7 | 680 | |||
GenBank mRNA | AK014519.1 | - | 7 | 679 | ||
AK021020.1 | - | 7 | 674 | |||
AK029084.1 | - | 7 | 646 | |||
AK041086.1 | - | 7 | 657 | |||
BC028754.1 | - | 7 | 635 | |||
BC086691.1 | - | 7 | 609 | 30682686 | ||
BC092523.1 | - | 7 | 622 | 30650002 | ||
U02601.1 | - | 7 | 662 | |||
U02602.1 | - | 7 | 662 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
