siRNA Details
| Gene Symbol: Tspyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009433.3 | NP_033459.1 | 1 | 1097 | ||
| GenBank mRNA | AF042180.1 | - | 1 | 1061 | ||
| AK053812.1 | - | 1 | 1048 | |||
| AK159432.1 | - | 1 | 1078 | |||
| AK166438.1 | - | 1 | 1078 | |||
| AK166805.1 | - | 1 | 1026 | |||
| AK166893.1 | - | 1 | 1026 | |||
| BC011213.1 | - | 1 | 1044 | 4196841 | ||

Tspyl1
Genomic Map