siRNA Details
| Gene Symbol: ITGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144996.1 | NP_001138468.1 | 14, 15 | 2244 | ||
| NM_001144997.1 | NP_001138469.1 | 14, 15 | 1769 | |||
| NM_002206.2 | NP_002197.2 | 14, 15 | 2232 | |||
| XM_005268839.1 | XP_005268896.1 | 15, 16 | 2497 | |||
| XM_005268840.1 | XP_005268897.1 | 15, 16 | 2479 | |||
| XM_005268841.1 | XP_005268898.1 | 15, 16 | 2497 | |||
| XM_005268842.1 | XP_005268899.1 | 14, 15 | 2347 | |||
| XM_005268844.1 | XP_005268901.1 | 16, 17 | 2122 | |||
| XM_005268846.1 | XP_005268903.1 | 15, 16 | 1903 | |||
| XM_005268848.1 | XP_005268905.1 | 14, 15 | 1921 | |||
| XM_005268849.1 | XP_005268906.1 | 14, 15 | 1851 | |||
| XM_005268850.1 | XP_005268907.1 | 12, 13 | 1740 | |||
| XM_017019265.1 | XP_016874754.1 | 13, 14 | 1825 | |||
| GenBank mRNA | AB210036.1 | - | 14, 15 | 2202 | ||
| AF032108.1 | - | 14, 15 | 2173 | |||
| AF052050.1 | - | 14, 15 | 2012 | |||
| AF072132.1 | - | 14, 15 | 2012 | |||
| AK022548.1 | - | 12, 13 | 1616 | |||
| AK304864.1 | - | 14, 15 | 1745 | |||
| AY358882.1 | - | 14, 15 | 2043 | |||
| BC050280.1 | - | 14, 15 | 2228 | 5284443 | ||

ITGA7
Genomic Map