siRNA Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077591.1 | NP_001071059.1 | 4 | 560 | ||
| NM_011651.2 | NP_035781.2 | 4 | 560 | |||
| GenBank mRNA | AF025310.2 | - | 4 | 540 | ||
| BC118618.1 | - | 4 | 539 | 40107271 | ||
| BC121827.1 | - | 4 | 539 | 40107270 | ||
| CN831975.1 | - | 283 | ||||

Tsks
Genomic Map