siRNA Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009441.2 | NP_033467.2 | 28 | 2898 | ||
| GenBank mRNA | AB008516.1 | - | 27 | 2775 | ||
| AK088273.1 | - | 27 | 2845 | |||
| AK163496.1 | - | 2 | 203 | |||
| AK165850.1 | - | 17 | 1806 | |||
| AK166120.1 | - | 18 | 1838 | |||
| AK220495.1 | - | 28 | 2940 | |||
| BC019173.1 | - | 28 | 2867 | |||
| BC037654.1 | - | 27 | 2798 | |||
| BC057207.1 | - | 27 | 2802 | |||
| BC145489.1 | - | 23 | 2418 | |||

Ttc3
Genomic Map