siRNA Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009441.2 | NP_033467.2 | 32 | 3405 | ||
| GenBank mRNA | AB008516.1 | - | 31 | 3282 | ||
| AK148494.1 | - | 5 | 429 | |||
| AK163496.1 | - | 6 | 710 | |||
| AK165850.1 | - | 21 | 2313 | |||
| AK220495.1 | - | 32 | 3447 | |||
| BC019173.1 | - | 32 | 3371 | |||
| BC037654.1 | - | 31 | 3305 | |||
| BC057207.1 | - | 31 | 3309 | |||
| BC145489.1 | - | 27 | 2925 | |||

Ttc3
Genomic Map