siRNA Details
| Gene Symbol: Tgoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009443.3 | NP_033469.1 | 2 | 881 | ||
| GenBank mRNA | AK041302.1 | - | 2 | 631 | ||
| AK076586.1 | - | 2 | 882 | |||
| AK133559.1 | - | 2 | 629 | |||
| AK161242.1 | - | 2 | 614 | |||
| AK162574.1 | - | 2 | 631 | |||
| BC009143.1 | - | 2 | 616 | 3157708 | ||
| BC141218.1 | - | 2 | 626 | 9055830 | ||
| BC145542.1 | - | 2 | 626 | 9054105 | ||
| CT010269.1 | - | 2 | 603 | |||
| D50031.1 | - | 2 | 629 | |||

Tgoln1
Genomic Map