siRNA Details
| Gene Symbol: Ttk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110265.1 | NP_001103735.1 | 2 | 197 | ||
| NM_001284272.1 | NP_001271201.1 | 2 | 197 | |||
| NM_009445.2 | NP_033471.2 | 2 | 197 | |||
| GenBank mRNA | AK041487.1 | - | 2 | 168 | ||
| AK049200.1 | - | 2 | 192 | |||
| AK084241.1 | - | 2 | 155 | |||
| AK164093.1 | - | 2 | 196 | |||
| AX775731.1 | - | 2 | 163 | |||
| BC058851.1 | - | 2 | 183 | 30023533 | ||
| BY729362.1 | - | 2 | 197 | |||
| M86377.1 | - | 2 | 145 | |||

Ttk
Genomic Map