siRNA Details
| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011652.3 | NP_035782.3 | 29 | 6870 | ||
| NM_028004.2 | NP_082280.2 | 28 | 6732 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttn| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011652.3 | NP_035782.3 | 29 | 6870 | ||
| NM_028004.2 | NP_082280.2 | 28 | 6732 | |||